missing translation for 'onlineSavingsMsg'
Learn More

Brady™ HandiMark™ Indoor/Outdoor Vinyl Tape

Brand:  Brady™ 142299

Code : 50

Additional Details : Weight : 0.02950kg

Product Code. 12810570

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Acrylic, Permanent, Cold Temperature
Purple
Vinyl
For facility and safety identification
60° 50 Gardner Units
-40°C to +82°C
15 m
HandiMark Indoor/Outdoor Vinyl Tape
Powder-coated
Gloss
HandiMark Indoor/Outdoor Tape
HandiMark Printer
1500/Pk.
0.024 mm
13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers