missing translation for 'onlineSavingsMsg'
Learn More
Learn More
H-B Instrument™ Durac™ Ungraduated Thermometers
For use in experiments demonstrating the construction of a thermometer, determining two fixed points and the differences between Celsius and Fahrenheit scales
Brand: H-B Instrument™ B62009-0100
This item is not returnable.
View return policy
Description
- Ungraduated—Students can mark their own graduations
- Organic liquid filled
- White back glass
- Individually packaged
Specifications
Experiments demonstrating the construction of a thermometer, determining two fixed points and the differences between Celsius and Fahrenheit scales | |
305 mm | |
Organic | |
110°C | |
-20°C | |
-20°C to +110°C |
12 in. | |
Glass | |
230°F | |
0°F | |
0°F to 230°F |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- H-B DURAC LIQUID-IN-GLASS UNGRADUATED THERMOMETER;
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use CO2, powder or waterspray to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : acetone