missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 120 Fast Filtration Sugar Beet Special Filter Paper

Used for analysis of juice in saccharimetric chains

£42.85

Specifications

For Use With (Application) Analysis of saccharine content
Format Cone shaped disc, pleated
Quantity 250/Pk.
Grade 120
View More Specs

Products 1
Product Code Brand Format Diameter (Metric) Disc Price Quantity & Availability  
Product Code Brand Format Diameter (Metric) Disc Price Quantity & Availability  
11794566
View Documents
Fisherbrand™
Cone shaped disc, pleated 210 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

  • Smooth surface filter paper 64g/m2
  • Used for analysis of juice in saccharimetric chains
Specifications

Specifications

Analysis of saccharine content
Cone shaped disc, pleated
250/Pk.
120
SDS