missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 1106 Cellulose Technical Filter Paper

Used for viscous liquids and coarse particles.

Brand:  Fisherbrand™

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11764464

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Thick smooth paper with large pore structure
Specifications

Specifications

Disc
50 to 80 μm
For syrups, animal fats, vegetable fats, margarines, Gelatin, dense oils, emulsions, essential oils
100 Pack
150 mm
Cellulose
50 to 80 μm
1106
0.47 mm
150 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers