missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 1105 Cellulose Technical Filter Paper

Suitable for filtration of large volume of liquid.

POA

Specifications

Format Disc
Material Cellulose
Particle Retention 3 to 6 μm
Pore Size 3 to 6 μm
For Use With (Application) For dyes, Ether, Alcoholic solutions, Eaux de Cologne, salted solutions, vegetal extracts, hair solutions, perfumes, tinctures, saccharine juices, vinegar
View More Specs

Description

Description

  • Very thick smooth paper
  • Wet strengthened
  • With high separation capacity
Specifications

Specifications

Disc
3 to 6 μm
For dyes, Ether, Alcoholic solutions, Eaux de Cologne, salted solutions, vegetal extracts, hair solutions, perfumes, tinctures, saccharine juices, vinegar
100 Pack
Cellulose
3 to 6 μm
1105
0.39 mm
Videos
SDS
Documents

Documents

Product Certifications