missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Grade 1103 Cellulose Technical Filter Paper

Suitable for removal of fine particles

Brand:  Fisherbrand™

Code : 47

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11759995

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Thick smooth paper
  • Wet strengthened
Specifications

Specifications

Disc
12 to 16 μm
For dyes, Ether, Alcoholic solutions, Eaux de Cologne, salted solutions, vegetal extracts, hair solutions, perfumes, tinctures, saccharine juices, vinegar
100 Pack
125 mm
Cellulose
12 to 16 μm
1103
0.3 mm
12 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers