missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR107 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25076-1-AP-20UL
This item is not returnable.
View return policy
Description
GPR107 also named as KIAA1624 or LUSTR1 is 600 amino acid protein, which belongs to the LU7TM family. GPR107 localizes in membrane and is a multi-pass membrane protein. GPR107 is a promising candidate receptor for neuronostatin, and neuronostatin, interacting with GPR107, may play an important role in the central control of cardiovascular function.Specifications
GPR107 | |
Polyclonal | |
Unconjugated | |
GPR107 | |
G protein coupled receptor 107, GCDRP, GPR107, KIAA1624, LUSTR1, Protein GPR107, RP11 88G17 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
277463, 57720 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5VW38, Q8BUV8 | |
Gpr107 | |
GPR107 Fusion Protein Ag18649 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |