missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva miniVE Vertical Electrophoresis System
Performs 1-D and 2-D electrophoresis and electrotransfer in single compact unit
Brand: Cytiva 80-6418-77
Code : GE
Additional Details : Weight : 2.39900kg
Includes: Three rectangular glass plates, three notched plates, two gel modules, lid, lower buffer chamber, Two each 1mm thick 10 well combs and 1mm thick spacer sets.
Description
- Performs two second-dimension separations using 7cm IPG strips in 1.5 hr.
- Runs two mini-gels or four mini-blots at once, using 10 × 10.5cm gel cassettes for increased separation distances
- Completes semi-wet blotting in 45 min. with only 300mL buffer
- Casts, runs, and blots mini-gels in four streamlined components: gel module, blot module, lower buffer chamber and safety lid
- Modular
- Uniform gel temperature is maintained with tank-style lower buffer chamber
- Full tank stabilizes temperature so that lanes are straight and bands are flat
- Blot Module will accept two gel/membrane stacks simultaneously
- Lower buffer chamber is designed to hold two Blot Modules, allowing four gels to be blotted at once
- Gel module is simultaneously a casting stand and an upper buffer chamber with integrated clamps
- Assembles gel sandwich of plates and spacers, clamps it onto upper buffer chamber, then pivot sealing gasket and cast gel
- After polymerization, release bottom seal and drop assembly into tank for running
- Handles gel cassettes up to 10 × 10.5cm (W × H), which increases separation length by more than 30% over conventional 10 × 8 cm gels
- Compatible with glass plates, combs, and spacers of SE 260 Mini-Vertical units
- For gradient gels, mini-Vertical Gel Casters can produce up to four identical gels at one time
- EPS 301 power supply
- 300V, 400 mAmp
- 600VAC (electrophoresis)
- For faster transfers use EPS 2A200
- Operating temperature: 5° to 75°C
- Tank volume: 1.7L with one module in place and 1.2L with two modules in place
Order Info
Weight: 1.15kg (2.53 lb.)
Specifications
EN61010-1, CE 73/23/EEC (LV directive) | |
Vertical Electrophoresis System | |
3 Rectangular Glass plates (10 x 10.5), 3 notched plates, 2 gel modules (8 x 9.5), lid, lower Buffer chamber, 2 each 1mm thick 10-well combs and 1mm thick spacer sets | |
1 to 2 |