missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCGR Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26784-1-AP-150UL
This item is not returnable.
View return policy
Description
Glucagon receptors are G-protein coupled receptors for glucagon and play a central role in the regulation of blood glucose levels and glucose homeostatsis. It regulates the rate of hepatic glucose production by promoting glycogen hydrolysis and gluconeogenesis. Glucagon receptors also play an important role in mediating the responses to fasting. Ligand binding causes a conformation change that triggers signaling via guanine nucleotide-binding proteins (G-proteins) and modulates the activity of down-stream effectors.Specifications
GCGR | |
Polyclonal | |
Unconjugated | |
GCGR | |
GCGR, GGR, GL R, glucagon receptor | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14527, 2642 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P47871, Q61606 | |
Gcgr | |
GCGR Fusion Protein Ag24861 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |