Learn More
3M™ Gas and Vapor Cartridge, 6000 Series
Combines excellent protection with superb comfort and balance. The 3M Gas and Vapor Cartridge is trapezoidal shaped and works with half- and full-facemasks that have bayonet connections systems.
Brand:  3M™ 7000034749
Code : NEW
Additional Details : Weight : 0.76050kg
Product Code. 15779007
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Each filter has a colored band using the EN14387 classification system, to denote the nature of the protection it provides.
  • Trapezoidal-shaped gas and vapor filters combine excellent protection with superb comfort and balance
  • Lightweight, low breathing resistance, and well balanced when fitted to appropriate mask
  • Provides excellent field of vision as a result of the unique trapezoidal shape.
  • Filters can be used on 3M™ Facemasks fitted with a bayonet connections system