Learn More
Fisherbrand™ Protective Disposable Face Masks
Provides convenient protection
Brand:  Fisherbrand™
Code : NEW
Additional Details : Weight : 2.21840kg
 View more versions of this product
Product Code. 15481932
  • £249.36 / Pack of 500
Estimated Shipment: 26-07-2021
Add to basket
You may only order up to the available stock. For more details call customer services on 01509 555 500
Description
Description
Provides convenient protection
  • Advanced protection
  • Disposable, single use
  • Choice of ear loops or tie closure
  • Nonwoven polypropylene outer layer
  • Thermally bonded nonwoven polypropylene inner layer
  • Latex-free
  • Choice of blue or white
  • Universal size/fit
Specifications
Specifications
Yes
3-ply
Non-woven Polypropylene (outer and inner), Meltblown Polypropylene (filter)
Polypropylene
Standard
Yes
Blue
Tie closures
Facemask