Learn More
Fisherbrand™ Protective Disposable Face Masks
Provides convenient protection
Brand:  Fisherbrand™
Code : NEW
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15451932
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Provides convenient protection
  • Advanced protection
  • Disposable, single use
  • Choice of ear loops or tie closure
  • Nonwoven polypropylene outer layer
  • Thermally bonded nonwoven polypropylene inner layer
  • Latex-free
  • Choice of blue or white
  • Universal size/fit
Specifications
Specifications
Yes
2-ply
Non-woven Polypropylene (outer), Thermally Bonded Non-woven Polypropylene (inner)
Polypropylene
Standard
Yes
White
Ear loops
Facemask