missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibronectin, MP Biomedicals™
Click to view available options
Quantity:
1 mg
Description
Human fibronectin (HFN) is suitable for use as an attachment factor in the propagation of cells in vitro when used to coat cell culture surfaces, including plasticware, glassware, and microcarrier beads. It is used in cell attachment and proliferation assays on human endothelial cells, human keratinocytes and human dermal fibroblasts.
Specifications
CAS | 86088-83-7 |
Quantity | 1 mg |
Synonym | CIG, Cold insoluble globulin, Fibronectin from human plasma= |
Solubility Information | Soluble in water (1mg/mL at 37°C). |
Percent Purity | ≥95% |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'