missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Taq DNA Polymerase, recombinant (5 U/μL)

Optimize routine PCR with this highly thermostable DNA polymerase.

Brand:  Thermo Scientific™ EP0403

Code : 88

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10541061

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Optimize routine PCR with this highly thermostable DNA polymerase.

Thermo Scientific ™ Taq DNA Polymerase is a highly thermostable DNA polymerase from the thermophilic bacterium Thermus aquaticus. The enzyme catalyzes 5'→3' synthesis of DNA, has no detectable 3'→5' exonuclease (proofreading) activity and possesses low 5'→3' exonuclease activity. In addition, Taq DNA Polymerase exhibits deoxynucleotidyl transferase activity, which frequently results in the addition of extra adenines at the 3'-end of PCR products. Recombinant Taq DNA Polymerase is the ideal tool for standard PCR of templates 5 kb or shorter.

PCR Master Mix is a 2X concentrated solution of Taq DNA Polymerase, dNTPs, and all of the components required for PCR, except DNA template and primers. This pre-mixed formulation saves time and reduces contamination due to a reduced number of pipetting steps required for PCR set up. The mix is optimized for efficient and reproducible PCR.

Highlights

  • Thermostable -half life is more than 40 min at 95°C
  • Generates PCR products with 3'-dA overhangs
  • Supplied with two buffers -10X Taq Buffer with KCl and 10X Taq Buffer with (NH4)2SO4. The latter allows for PCR at wide range of magnesium concentrations and decreases unspecific priming
  • Incorporates modified nucleotides (e.g., biotin-, digoxigenin-, fluorescently-labeled nucleotides)
Specifications

Specifications

2 x 1.25 mL 10X Taq Buffer with (NH4)2SO2, 2 x 1.25 mL 10X Taq Buffer with KCl 25 and 2 x 1.25 mLmM MgCl2
100 Units
Half life is more than 40 min. at 95°C
Taq DNA Polymerase (recombinant), 1 U/μL
Standard PCR, DNA Sequencing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.