Learn More
Thermo Scientific™ F1-ClipTip™ Multichannel Pipettes
GREENER_CHOICE
Ensure a complete seal on every channel with minimal tip attachment and ejection force using these multichannel pipetters, which feature an innovative locking technology.
Marca:  Thermo Scientific™ 4661120
 vedi altre versioni di questo prodotto
Codice prodotto. 13236299
Articolo fuori commercio e non più disponibile. Visualizzare il prodotto per possibili alternative o chiedere assistenza alla nostra divisione di supporto tecnoco telefonando al n. 056 618 41 11.
Visualizzare prodotti alternativi »
Esplora altre offerte speciali
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Descrizione
Descrizione
Ensure a complete seal on every channel with minimal tip attachment and ejection force using Thermo Scientific™ F1-ClipTip™ Pipettes, which feature innovative ClipTip interlocking technology. Once attached, tips are locked firmly in place and will not loosen or fall off regardless of application pressure. The innovative three interlocking clip design ensures the tip is held securely on the F1-ClipTip pipette until it is released.
Features
  • Tip is locked and sealed in place until ejected—no dropped tips that waste valuable samples
  • Complete seal to secure the sample volume in each tip enhanced accuracy and precision
  • Tips are attached and ejected with a light touch—eliminates the frustration of banging tips on the pipette
Ergonomic Design
  • Offering low attachment, ejection and plunger forces for safe and comfortable use
Remarkably Lightweight
  • Modern design protects user from the risk of Repetitive Strain Injuries (RSI)
Adjustable Comfort
  • The 120° adjustable finger rest—for both right- and left-handed operator comfort—allows the hand to relax during pipetting cycles
Super Blow-out
  • 150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action†
Soft-touch Tip Ejection
  • Innovative gearing mechanism produces more downward force with less effort, releasing tips with a gentle pus
Volume Lock Adjustment
  • The secure volume setting prevents undesirable volume drift during operation
Industry-leading Lifetime Warranty
  • With convenient online registration
† Available in 50μL models and below
TRUSTED_SUSTAINABILITY
Specifica
Specifica
Da 1.5 a 5%
F1-ClipTip 50
F1-ClipTip™ 50
8
Pipette
0.1 µl
Il sistema di pipettaggio F1-ClipTip è provvisto di marchio CE in conformità alla direttiva IVD in Europa. Prodotto in conformità a ISO 9001, ISO 14001 e ISO 13485.
Viola
F1-ClipTip™
F1-ClipTip™
Da 5 a 50 µl
Da 0.7 a 2%
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documenti
Documenti
Certificati
Vedi i prodotti accessori
Offerte speciali
Offerte speciali