Learn More
Eppendorf™ ep Dualfilter T.I.P.S.™ SealMax Filter Pipet Tips
Tips with liquid blocking technology provide additional protection against over pipetting
POA
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
Format | Rack |
Material | PE (Polyethylene) |
Description
- Sterile tips made of polyethylene
- White and violet filter layers in the tip retain aerosols and biomolecules
- Purple filter layer blocks liquids, binds aerosols and does not allow any liquid to pass through
- Hydrophobic white layer repels drops, splashes and binds aerosols
- Free of PCR inhibiting additives
- Provide protection against contamination of pipet and sample
- Color coded tip boxes simplify matching pipetes with their tips
PCR and microbiology procedures
Order Info
10 Racks x 96 tips
Specifications
Filtered | |
Sterile | |
Rack | |
PE (Polyethylene) |