Learn More
3M™ Petrifilm™ Environmental Listeria Spreader 6498
Spreader used to distribute the sample evenly for 3M™ Petrifilm Environmental Listeria Plates.
Brand: 3M™ Petrifilm™ 7100067523
Additional Details : Weight : 0.00100kg
Description
- Large square spreader for 3M™ Petrifilm Environmental Listeria Plates.
- Spreads inoculum over uniform area.
- Clear acrylic material allows the technician to observe and control spread.
Specifications
Accessory | |
2 Pk. | |
3M™ Petrifilm Environmental Listeria Plates |