Learn More
Thermo Scientific™ Click-On™ Inline Gas Filter Connectors
Use these quality end fittings o-rings, and other connectors for your Thermo Scientific™ Click-On™ Inline Filters.
Brand:  Thermo Scientific™ 60180-813
Code : 16
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10117844
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Use these quality end fittings o-rings, and other connectors for your Thermo Scientific™ Click-On™ Inline Filters. The end fittings remain installed during filter replacement, thereby reducing the cost of clean gas.
  • Incorporates needle valves that close lines and prevent contamination
  • End fittings measure 0.125in. (0.3cm) or 0.25in. (0.6cm)
  • Reusable
Specifications
Specifications
Click-On Inline Filter
N/A
N/A
Double Ended Connector to Connect Filter to Indicator
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.