Learn More
Merck Millipore PetriSlide™ Dishes Preloaded with Absorbent Pads
Substitute for Petri Dishes. PetriSlide™ Dish Preloaded with Absorbent Pads is designed to hold filter securely in place. Transparent cover allows microscopic examination without removal. Rectangular base has rounded corners for mounting on microscope stage.
Brand:  Merck Millipore PDMA04700
Code : 71
Additional Details : Weight : 1.13850kg
Product Code. 10122104
  • £135.00 / Pack of 100
In stock, ready to ship: 8
Add to basket
Description
Description
Substitute for Petri Dishes. PetriSlide™ Dish Preloaded with Absorbent Pads is designed to hold filter securely in place. Transparent cover allows microscopic examination without removal. Rectangular base has rounded corners for mounting on microscope stage.
Key Applications:
  • Clinical Lab
  • Food and Beverage
  • Industrial QC
  • Environmental Analysis
Specifications
Specifications
Clear
47 mm
Polystyrene
For microbiological analysis
Round
Sterile
Petri Dish