missing translation for 'onlineSavingsMsg'
Learn More

Merck MF-Millipore™ Membrane Filter, 0.1 μm

13 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic

Brand:  Merck VCWP09025

Code : 71

Additional Details : Weight : 0.01000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II


 View more versions of this product

Product Code. 10450971

  • £285.00 / Pack of 25
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications.

MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.

Features & Benefits:

  • Versatile filter for biological and environmental monitoring applications
  • Available in a range of pore sizes, colored black or white, with or without a gridded surface
  • Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Specifications

Specifications

Not Autoclavable
MCE
Non-sterile
105 μm
Hydrophilic
90 mm
White
0.1 μm
1.5 mL/min x cm2
Filter Membranes
90 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MCE 0.1µm WH PL 90mm

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use powder to extinguish.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose, octylphenol polyethoxyethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers