missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF4EBP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11182-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the eukaryotic translation initiation factor 4E binding protein family. The gene products of this family bind eIF4E and inhibit translation initiation. However, insulin and other growth factors can release this inhibition via a phosphorylation-dependent disruption of their binding to eIF4E. Regulation of protein production through these gene products have been implicated in cell proliferation, cell differentiation and viral infection.Specifications
EIF4EBP2 | |
Polyclonal | |
Unconjugated | |
EIF4EBP2 | |
4E BP2, 4EBP2, eIF4E binding protein 2, EIF4EBP2, PHASII | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13688, 1979 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P70445, Q13542 | |
EIF4EBP2 | |
EIF4EBP2 Fusion Protein Ag1670 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |