missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ ECO AluRack for Upright Freezers, 100 mm Box Height
ECO AluRack fits into any upright freezer and can be used with all standard cryoboxes.
£67.95 - £87.95
Specifications
Description | Seawater resistant |
---|---|
Material | Aluminum |
For Use With (Equipment) | Upright Freezers |
Height (Metric) | 100 mm (Box) |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
12402560
|
Tenak™
TE11310 |
Boxes (10) |
£79.15
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
12412550
|
Tenak™
TE11294 |
Boxes (6) |
£67.95
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
12422560
|
Tenak™
TE11312 |
Boxes (15) |
£87.95
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
12432550
|
Tenak™
TE11296 |
Boxes (9) |
£72.80
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
12462550
|
Tenak™
TE11302 |
Boxes (8) |
£72.80
Each |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
12482550
|
Tenak™
TE11304 |
Boxes (12) |
£79.15
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
Description
- Made of seawater resistant aluminum
- Fits into any freezer and can be used with all standard cryoboxes
- Racks can be used with all freezer brands
- Without white cardboard boxes
- Holds is expressed by Height x Dept
Specifications
Seawater resistant | |
Aluminum | |
Upright Freezers | |
100 mm (Box) |