missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dystroglycan Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2B1G12, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
Antigen | Dystroglycan |
---|---|
Clone | 2B1G12 |
Concentration | 1 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16808895
|
Proteintech
66735-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16898885
|
Proteintech
66735-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dystroglycan is a laminin binding component of the dystrophin-glycoprotein complex which provides a linkage between the subsarcolemmal cytoskeleton and the extracellular matrix. Dystroglycan 1 is a candidate gene for the site of the mutation in autosomal recessive muscular dystrophies. The dramatic reduction of dystroglycan 1 in Duchenne muscular dystrophy leads to a loss of linkage between the sarcolemma and extracellular matrix, rendering muscle fibers more susceptible to necrosis. Dystroglycan also functions as dual receptor for agrin and laminin-2 in the Schwann cell membrane. The muscle and nonmuscle isoforms of dystroglycan differ by carbohydrate moieties but not protein sequence. Alternative splicing results in multiple transcript variants all encoding the same protein.Specifications
Dystroglycan | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
156DAG, A3a, AGRNR, Alpha DG Beta dystroglycan, Beta DG, DAG, DAG1, Dystroglycan | |
Dag1 | |
IgG1 | |
Protein G | |
Antibody |
2B1G12 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Rat, Porcine, Mouse, Human | |
Q14118, Q29243, Q62165 | |
100620392, 114489, 13138, 1605 | |
Dystroglycan Fusion Protein Ag27222 | |
Primary | |
-20°C | |
DAG1 |