missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Dosing/Measuring Spoon Set
BIOPHARMA

Spoon set can be used to precisely measure and dose small amounts of powder, granules, and liquids.

Brand:  Buerkle™ 5378-4125

9.20 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 16535260

  • £12.20 / 1 set
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For precise measuring, excess medium is stripped off the upper edge of the spoon
  • The content volume is engraved on the handle
  • The flat front edge of the volumetric spoon is ideal for picking up medium from the bottom of the container or for getting into the corners of the container, allowing for good emptying of small remaining amounts
  • The flat base makes it possible to set the volumetric spoon down without spilling the content
  • An eyelet in the end of the handle can be used to hang up a spoon or to bundle several together
  • Content volume: 0.5, 1.0, 2.5, 5.0, 10, 15, 25, and 50 mL
  • Set of volumetric spoons with eight sizes
  • With stripper edge
  • Complies with EU food requirements and FDA regulations
Specifications

Specifications

Sampler
Dosing spoons with stripper edge for exact measuring
Single-use
Blue
Especially for The Food, Animal Feed and Pharmaceutical Industries
Polystyrene
0.5 to 50 mL
Non-sterile
Food Contact, Reach, RoHS
100
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers