missing translation for 'onlineSavingsMsg'
Learn More

Dodecacarbonyltriiron, 96% (dry wt.), stab. with 5-10% methanol, Thermo Scientific Chemicals

Catalog Number p-7049152
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
17685-52-8
MFCD00135617
triiron dodecarbonyl
This item is not returnable. View return policy
17685-52-8
MFCD00135617
triiron dodecarbonyl

Reducing agent (reduces NO2), catalyst for alkyne trimerization

It eliminates sulfur from substituted thiophene complexes, which leads to ferroles. Access to new areas of organometallic chemistry is promising using the reaction of Fe3(CO)12 with tellurium-nitrogen heterocycles such as benzoisotellurazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 140°C (decomposition)
Color Black
Density 2.00 g/mL
Linear Formula Fe3(CO)12
Quantity 2 g
UN Number UN3179
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water. Soluble in non polar organic solvents.
Formula Weight 503.67
Percent Purity 96%
Physical Form Crystalline
Assay (dry wt.)
Chemical Name or Material Dodecacarbonyltriiron, Stabilized with 5 to 10% methanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dodecacarbonyltriiron, stab. with 5-10% methanol
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 2
Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H371-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Dodecacarbonyltriiron, Methanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'