Learn More
DNPEP Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15016-1-AP-150UL
 View more versions of this product
Product Code. 16853984
  • £349.00 / 150µL
Explore more special offers
Description
Description
Aminopeptidase with specificity towards an acidic amino acid at the N-terminus. Likely to play an important role in intracellular protein and peptide metabolism.
Specifications
Specifications
DNPEP
Polyclonal
Unconjugated
DNPEP
aspartyl aminopeptidase, ASPEP, DAP, DNPEP
Rabbit
Antigen Affinity Chromatography
RUO
23549
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9ULA0
DNPEP
DNPEP Fusion Protein Ag6966
150 μL