missing translation for 'onlineSavingsMsg'
Learn More

Divinyltin dichloride, Thermo Scientific™

£22.00 - £335.00

Chemical Identifiers

CAS 7532-85-6
Molecular Formula C4H8Cl2Sn
Molecular Weight (g/mol) 245.72
MDL Number MFCD00058728
InChI Key OFJXSWMLVIFLKC-UHFFFAOYSA-N
Synonym divinyltin dichloride, dichlorodivinylstannane, stannane, dichlorodiethenyl, dichloro divinyl stannane, divinyltindichloride, acmc-20al40, dichloro-bis ethenyl stannane
PubChem CID 82033
IUPAC Name dichloro-bis(ethenyl)stannane
SMILES Cl.Cl.C=C[SnH2]C=C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11374547
View Documents
Thermo Scientific Alfa Aesar
039613.04
2 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11354547
View Documents
Thermo Scientific Alfa Aesar
039613.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11364547
View Documents
Thermo Scientific Alfa Aesar
039613.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

7532-85-6
245.72
OFJXSWMLVIFLKC-UHFFFAOYSA-N
82033
Cl.Cl.C=C[SnH2]C=C
C4H8Cl2Sn
MFCD00058728
divinyltin dichloride, dichlorodivinylstannane, stannane, dichlorodiethenyl, dichloro divinyl stannane, divinyltindichloride, acmc-20al40, dichloro-bis ethenyl stannane
dichloro-bis(ethenyl)stannane

Specifications

7532-85-6
C4H8Cl2Sn
UN2788
divinyltin dichloride, dichlorodivinylstannane, stannane, dichlorodiethenyl, dichloro divinyl stannane, divinyltindichloride, acmc-20al40, dichloro-bis ethenyl stannane
Cl.Cl.C=C[SnH2]C=C
245.72
243.69
Divinyltin dichloride
54°C to 56°C (3 mmHg)
MFCD00058728
Moisture sensitive
OFJXSWMLVIFLKC-UHFFFAOYSA-N
dichloro-bis(ethenyl)stannane
82033
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c

H301+H311-H330

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

missing translation for 'einecsNumber' : 231-399-1

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications