missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyldichlorosilane, Pure, Fisher Chemical™
Click to view available options
Quantity:
100 mL
250 mL
Specifications
Melting Point | -76°C |
Boiling Point | 70°C |
Packaging | Plastic Coated Glass bottle |
Quantity | 100 mL |
Formula Weight | 129.06g/mol |
Vapor Pressure | 150 hPa at 20°C |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Dichlorodimethylsilane
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 A
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'