missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Sulfoxide, for HPLC, Fisher Chemical™
83.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
83.31 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "22914" to get your promotional price.
83.31 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
250 mL
500 mL
2.5 L
Specifications
Melting Point | 18.4°C |
Color | Colorless |
Boiling Point | 189°C |
Quantity | 250 mL |
Packaging | Plastic-coated Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 0.55mbar at 20°C |
Viscosity | 1.98 mPaS at 25°C |
IUPAC Name | methanesulfinylmethane |
Formula Weight | 78.13g/mol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'