missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxymethylvinylsilane, 97%, Thermo Scientific Chemicals

Product Code. 11338915
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16753-62-1
Molecular Formula C5H12O2Si
Molecular Weight (g/mol) 132.234
MDL Number MFCD00129170
InChI Key ZLNAFSPCNATQPQ-UHFFFAOYSA-N
Synonym vinylmethyldimethoxysilane, dimethoxymethylvinylsilane, dimethoxy methyl vinyl silane, silane, ethenyldimethoxymethyl, ethenyldimethoxymethylsilane, methyldimethoxyvinylsilane, vinyl methyldimethoxysilane, dimethoxy methyl vinylsilane #, methylvinyldimethoxysilane
PubChem CID 85589
IUPAC Name ethenyl-dimethoxy-methylsilane
SMILES CO[Si](C)(C=C)OC

Specifications

Boiling Point 102°C to 104°C
Quantity 5 g
UN Number UN1993
Beilstein 3043765
Flash Point 5°C (41°F)
Formula Weight 132.23
Percent Purity 97%
Refractive Index 1.395
Sensitivity Moisture sensitive
Density 0.889
Chemical Name or Material Dimethoxymethylvinylsilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'