Learn More
Trajan Scientific and Medical™ Diluter/Dispenser Syringes for V6 Pumps
Gives better performance and superior quality. Trajan Scientific and Medical Diluter/Dispenser Syringes for V6 Pumps has the knowledge and expertise that goes into manufacturing an SGE syringe.
Brand:  Trajan Scientific and Medical™ 008996
 View more versions of this product
Product Code. 15278513
  • £140.00 / Each
Explore more special offers
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Diluter/Dispenser Syringe for V6 Pumps
Specifications
Specifications
60mm stroke, 1/4-28 UNF thread connection, 4.8mm hole in plunger
10 mL
V6 Pump