Learn More
Medline Scientific™ Digi Max II Trinocular Digital Camera Microscope
A dedicated digital camera microscope using a 3Mpx digital USB camera and advanced software to allow for image capture and analysis.
Brand:  Medline Scientific™ 1202.8000M
Additional Details : Weight : 10.00000kg
Product Code. 12737367
  • £2140.00 / Each
Estimated Shipment: 01-05-2024
to see stock.
Add to basket
Description
Description
  • SI 3000 camera system
  • Cost-effective option, perfect for under- and post-graduate teaching laboratories
  • Ideally suited to routine, benchtop microscopy work for general purpose slide examinations where prolonged usage is not the norm
  • Integrated carrying handle
  • Trinocular 30° head
  • 4x, 10x, 100x achromatic objectives
Specifications
Specifications
3Mpx digital USB
4X, 10X, 40X, 100X
USB cable
Trinocular, 30° inclined
Achromatic
Integral SI 3000