missing translation for 'onlineSavingsMsg'
Learn More

Dibenzyl phosphite, 90+%, Thermo Scientific Chemicals

Catalog Number p-7045218
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
17176-77-1
C14H14O3P
261.24
MFCD00004774
RQKYHDHLEMEVDR-UHFFFAOYSA-N
dibenzyl phosphonate, phosphonic acid dibenzyl ester, unii-1o720l5h5a, phosphonic acid, bis phenylmethyl ester, dibenzylphosphit, dibenzylphosphonate, phosphonic acid bis phenylmethyl ester, oxo-bis phenylmethoxy phosphanium, mibxhgzaarwagi-uhfffaoysa-n, dibenzyl phosphite, technical grade
6334615
oxo-bis(phenylmethoxy)phosphanium
O=[P+](OCC1=CC=CC=C1)OCC1=CC=CC=C1
This item is not returnable. View return policy
17176-77-1
C14H14O3P
261.24
MFCD00004774
RQKYHDHLEMEVDR-UHFFFAOYSA-N
dibenzyl phosphonate, phosphonic acid dibenzyl ester, unii-1o720l5h5a, phosphonic acid, bis phenylmethyl ester, dibenzylphosphit, dibenzylphosphonate, phosphonic acid bis phenylmethyl ester, oxo-bis phenylmethoxy phosphanium, mibxhgzaarwagi-uhfffaoysa-n, dibenzyl phosphite, technical grade
6334615
oxo-bis(phenylmethoxy)phosphanium
O=[P+](OCC1=CC=CC=C1)OCC1=CC=CC=C1

Dibenzyl phosphite is used for the phosphorylation of phenols involves reaction in the presence of N-ethyldiisopropylamine and DMAP to give the dibenzyl phenyl phosphate. It is used in the preparation of phenylalkylphosphonamidates which are then used as probes for a hydrophobic binding register in prostate-specific membrane antigen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dibenzyl phosphite is used for the phosphorylation of phenols involves reaction in the presence of N-ethyldiisopropylamine and DMAP to give the dibenzyl phenyl phosphate. It is used in the preparation of phenylalkylphosphonamidates which are then used as probes for a hydrophobic binding register in prostate-specific membrane antigen.

Solubility
Soluble in DMSO, Methanol. Reacts with water.

Notes
Store at 4°C. Moisture Sensitive. Incompatible with moisture and oxidizing agents. Store under dry inert gas. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.187
Boiling Point 110°C to 120°C (0.1 mmHg)
Flash Point >110°C (230°F)
Odor Odorless
Refractive Index 1.554
Quantity 5 g
Beilstein 1982794
Sensitivity Moisture sensitive
Merck Index 14,3014
Solubility Information Soluble in DMSO,Methanol. Reacts with water.
Formula Weight 262.25
Percent Purity ≥90%
Chemical Name or Material Dibenzyl phosphite
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'