missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEXI Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14811-1-AP-150UL
This item is not returnable.
View return policy
Description
DEXI (dexamethasone-induced protein), also known as MYLE, is a 95 amino acid protein belonging to the DEXI family.Specifications
DEXI | |
Polyclonal | |
Unconjugated | |
DEXI | |
Dexamethasone induced protein, DEXI, MYLE, Protein MYLE | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
28955, 497857, 58239 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95424, Q9WUQ7 | |
DEXI | |
DEXI Fusion Protein Ag6535 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |