missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP2S1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19100-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum. In rodents, the homologous protein has been shown to metabolize certain carcinogens; however, the specific function of the human protein has not been determined.Specifications
CYP2S1 | |
Polyclonal | |
Unconjugated | |
CYP2S1 | |
CYP2S1, CYPIIS1, Cytochrome P450 2S1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29785, 74134 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96SQ9, Q9DBX6 | |
Cyp2s1 | |
CYP2S1 Fusion Protein Ag4605 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |