missing translation for 'onlineSavingsMsg'
Learn More

α-Cyano-3-hydroxycinnamic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-94871
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
54673-07-3
C10H7NO3
189.17
MFCD00004203
HPLNTJVXWMJLNJ-YWEYNIOJSA-N
2z-2-cyano-3-3-hydroxyphenyl prop-2-enoic acid
21338270
(Z)-2-cyano-3-(3-hydroxyphenyl)prop-2-enoic acid
C1=CC(=CC(=C1)O)C=C(C#N)C(=O)O
This item is not returnable. View return policy
54673-07-3
C10H7NO3
189.17
MFCD00004203
HPLNTJVXWMJLNJ-YWEYNIOJSA-N
2z-2-cyano-3-3-hydroxyphenyl prop-2-enoic acid
21338270
(Z)-2-cyano-3-(3-hydroxyphenyl)prop-2-enoic acid
C1=CC(=CC(=C1)O)C=C(C#N)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 230.0°C to 233.0°C
Color Green-Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HOC6H4CH=C(CN)CO2H
Quantity 5 g
Beilstein 10, 520
Formula Weight 189.17
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material α-Cyano-3-hydroxycinnamic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • a-Cyano-3-hydroxycinnamic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'