missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin), 98+%

Catalog Number p-4510042
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
458-37-7
C21H20O6
368.39
MFCD00008365
ZIUSSTSXXLLKKK-KOBPDPAPSA-N
curcumin, diferuloylmethane, natural yellow 3, turmeric yellow, turmeric, curcuma, kacha haldi, gelbwurz, halad, curcumin i
969516
CHEBI:3962
COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
458-37-7
C21H20O6
368.39
MFCD00008365
ZIUSSTSXXLLKKK-KOBPDPAPSA-N
curcumin, diferuloylmethane, natural yellow 3, turmeric yellow, turmeric, curcuma, kacha haldi, gelbwurz, halad, curcumin i
969516
CHEBI:3962
COC1=CC(\C=C\C(\O)=C\C(=O)\C=C\C2=CC=C(O)C(OC)=C2)=CC=C1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 183.0°C
Color Orange
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula [HOC6H3(OCH3)CH=CHCO]2CH2
Refractive Index 1.4155 to 1.4175
Quantity 10 g
Beilstein 08,IV,3697
Merck Index 15,2663
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in diethyl ether,soluble in alcohol,glacial acetic acid,acetone
IUPAC Name (1E,6E)-1,7-bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione
Formula Weight 368.38
Percent Purity 98+%
Physical Form Fine Crystalline Powder
Chemical Name or Material Curcumin (mixture of curcumin, demethoxycurcumin, and bisdemethoxycurcumin)
Show More Show Less
compliance-icons
Product Identifier
  • Curcumin
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'