Learn More
Thermo Scientific™ Culti-Loops™ Rhodotorula glutinis ATCC™ 32765™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Brand: Thermo Scientific™ R4607095
191.75 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Specifications
32765TM* | |
Culti-Loops | |
Fungi | |
5 Loops/Package |
Rhodotorula glutinis ATCC™ 32765™ | |
Culti-Loops Quality Control Organism | |
Qualitative QC | |
CBS 6020 [CCRC 20576, MUCL 30684] |
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.