Learn More
Thermo Scientific™ Culti-Loops™ Lysinibacillus sphaericus ATCC™ 4525™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Brand:  Thermo Scientific™ R4601229
199.71 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Product Code. 12918868
  • £189.00 / Pack of 5
Explore more special offers
Description
Description
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lysinibacillus species
Lysinibacillus sphaericus ATCC™ 4525™
Culti-Loops Quality Control Organism
Qualitative QC
WDCM00139, 25 [CCM 2177, CIP 104788, NRS 348]
4525TM*
Culti-Loops
Gram-positive Rods
5 Loops/Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.