missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12600-1-AP-20UL
This item is not returnable.
View return policy
Description
Three different procarboxypeptidases A and two different procarboxypeptidases B have been isolated. The B1 and B2 forms differ from each other mainly in isoelectric point. Carboxypeptidase B1 is a highly tissue-specific protein and is a useful serum marker for acute pancreatitis and dysfunction of pancreatic transplants. It is not elevated in pancreatic carcinoma.Specifications
CPB1 | |
Polyclonal | |
Unconjugated | |
CPB1 | |
Carboxypeptidase B, carboxypeptidase B1 (tissue), CPB, CPB1, Pancreas specific protein, PASP, PCPB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1360, 24271, 76703 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P15086, P19223 | |
Cpb1 | |
CPB1 Fusion Protein Ag3243 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |