missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) sulfate pentahydrate, ≥98.0%, Reagent Grade, Honeywell™

Product Code. 15687770
Click to view available options
Quantity:
250 g
500 g
1 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade,≥98.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper;sulfate;pentahydrate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Melting Point 110°C
Color Blue Green
pH 3.5 to 4.5 (at 20°C)
Physical Form Solid
Quantity 250 g
Linear Formula CuSO4 · 5H2O
UN Number UN3077
Formula Weight 249.69g/mol
Percent Purity ≥98.0%
Odor Odorless
Packaging Poly bottle
Vapor Pressure 7.3mmHg (25°C)
Density 2.28g/cu.cm at 20°C
Chemical Name or Material Copper(II) sulfate pentahydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'