missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) sulfate pentahydrate, ≥98.0%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
250 g
500 g
1 kg
Packaging:
Poly bottle
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Melting Point | 110°C |
Color | Blue Green |
pH | 3.5 to 4.5 (at 20°C) |
Physical Form | Solid |
Quantity | 1 kg |
Linear Formula | CuSO4 · 5H2O |
UN Number | UN3077 |
Formula Weight | 249.69g/mol |
Percent Purity | ≥98.0% |
Odor | Odorless |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'