missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(I) Bromide, ≥98.0% (RT), Honeywell™
Purum p.a.,≥98.0% (RT)
£12.48 - £94.76
Chemical Identifiers
CAS | 7787-70-4 |
---|---|
Molecular Formula | BrCu |
Molecular Weight (g/mol) | 143.45 |
MDL Number | MFCD00010969 |
InChI Key | NKNDPYCGAZPOFS-UHFFFAOYSA-M |
Synonym | copper i bromide, copper bromide cubr, copper 1+ bromide, copper monobromide, cuprousbromide, hsdb 270, cubr, copper 1 bromide |
PubChem CID | 24593 |
SMILES | [Cu+].[Br-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15696010
|
Honeywell Research Chemicals
61163-10G |
10 g | Poly bottle | N/A | |||||
15686010
|
Honeywell Research Chemicals
61163-100G |
100 g | Poly bottle | N/A | |||||
15606020
|
Honeywell Research Chemicals
61163-500G |
500 g | Poly Bottle | N/A | |||||
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
7787-70-4 | |
143.45 | |
NKNDPYCGAZPOFS-UHFFFAOYSA-M | |
24593 |
BrCu | |
MFCD00010969 | |
copper i bromide, copper bromide cubr, copper 1+ bromide, copper monobromide, cuprousbromide, hsdb 270, cubr, copper 1 bromide | |
[Cu+].[Br-] |
Specifications
1345°C (1013hPa) | |
7787-70-4 | |
Powder | |
CuBr | |
NONH for all modes of transport | |
NKNDPYCGAZPOFS-UHFFFAOYSA-M | |
λ¹-copper(1+) bromide | |
24593 | |
≥98.0% (RT) | |
4.71g/mL at 25°C |
492°C | |
Colorless | |
BrCu | |
MFCD00010969 | |
copper i bromide, copper bromide cubr, copper 1+ bromide, copper monobromide, cuprousbromide, hsdb 270, cubr, copper 1 bromide | |
[Cu+].[Br-] | |
143.45 | |
143.45g/mol | |
Odorless | |
Copper(I) bromide |
Safety and Handling
missing translation for 'rtecsNumber' : 232-131-6