missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) Bromide, ≥98.0% (RT), Honeywell™

Product Code. 15686010
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Poly bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Purum p.a.,≥98.0% (RT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7787-70-4
Molecular Formula BrCu
Molecular Weight (g/mol) 143.45
MDL Number MFCD00010969
InChI Key NKNDPYCGAZPOFS-UHFFFAOYSA-M
Synonym copper i bromide, copper bromide cubr, copper 1+ bromide, copper monobromide, cuprousbromide, hsdb 270, cubr, copper 1 bromide
PubChem CID 24593
SMILES [Cu+].[Br-]

Specifications

Boiling Point 1345°C (1013hPa)
Melting Point 492°C
Color Colorless
Physical Form Powder
Quantity 100 g
Linear Formula CuBr
UN Number NONH for all modes of transport
IUPAC Name λ¹-copper(1+) bromide
Formula Weight 143.45g/mol
Percent Purity ≥98.0% (RT)
Odor Odorless
Packaging Poly bottle
Density 4.71g/mL at 25°C
Chemical Name or Material Copper(I) bromide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'