missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(I) Bromide, ≥98.0% (RT), Honeywell™
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Poly Bottle
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Specifications
Quantity | 500 g |
Linear Formula | CuBr |
UN Number | NONH for all modes of transport |
IUPAC Name | bromocopper |
Formula Weight | 143.45g/mol |
Percent Purity | ≥98.0% (RT) |
Packaging | Poly Bottle |
Density | 4.71g/mL at 25°C |
Chemical Name or Material | Copper(I) bromide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'