Learn More
Cobalt(II) gluconate hydrate, Thermo Scientific Chemicals
£29.25 - £91.05
Chemical Identifiers
CAS | 1809162-99-9 |
---|---|
Molecular Formula | C12H26CoO15 |
Molecular Weight (g/mol) | 469.258 |
MDL Number | MFCD02684006 |
InChI Key | QBVYGKYWWCNZSO-XRDLMGPZSA-N |
Synonym | cobalt ii gluconate hydrate, 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoyl oxy cobaltio 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoate hydrate |
PubChem CID | 117064710 |
IUPAC Name | cobalt;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid;hydrate |
SMILES | C(C(C(C(C(C(=O)O)O)O)O)O)O.C(C(C(C(C(C(=O)O)O)O)O)O)O.O.[Co] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11475707
|
Thermo Scientific Alfa Aesar
B24293.14 |
25 g |
£29.25
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11485707
|
Thermo Scientific Alfa Aesar
B24293.22 |
100 g |
£91.05
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1809162-99-9 | |
469.258 | |
QBVYGKYWWCNZSO-XRDLMGPZSA-N | |
117064710 | |
C(C(C(C(C(C(=O)O)O)O)O)O)O.C(C(C(C(C(C(=O)O)O)O)O)O)O.O.[Co] |
C12H26CoO15 | |
MFCD02684006 | |
cobalt ii gluconate hydrate, 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoyl oxy cobaltio 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoate hydrate | |
cobalt;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid;hydrate |
Specifications
>300°C | |
C12H26CoO15 | |
cobalt ii gluconate hydrate, 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoyl oxy cobaltio 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoate hydrate | |
C(C(C(C(C(C(=O)O)O)O)O)O)O.C(C(C(C(C(C(=O)O)O)O)O)O)O.O.[Co] | |
469.258 | |
449.23 (Anhydrous) |
1809162-99-9 | |
MFCD02684006 | |
QBVYGKYWWCNZSO-XRDLMGPZSA-N | |
cobalt;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid;hydrate | |
117064710 | |
Cobalt(II) gluconate hydrate |
Safety and Handling
GHS H Statement
H301-H332-H317
Toxic if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
P261-P264b-P270-P271-P272-P280g-P301+P312-P302+P352-P304+P340-P312-P330-P333+P313-P363-P501c
H302+H332-H317
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 276-206-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only