missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Classic Rack for Chest Freezers, 50 mm Box Height, with Boxes
Classic Rack is suitable for use with all chest freezer models.
£66.00 - £107.00
Specifications
Includes | White cardboard boxes, 9 x 9 cell dividers |
---|---|
Material | Stainless Steel |
For Use With (Equipment) | Chest Frseezers |
Height (Metric) | 50 mm (Box) |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
15200484
|
Tenak™
TE21440B |
Boxes (8) |
£87.45
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
15210484
|
Tenak™
TE21441B |
Boxes (9) |
£91.40
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
15230484
|
Tenak™
TE21448B |
Boxes (3) |
£66.00
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
15250484
|
Tenak™
TE21449B |
Boxes (5) |
£73.80
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
15260484
|
Tenak™
TE21450B |
Boxes (6) |
£77.70
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
15270484
|
Tenak™
TE21451B |
Boxes (7) |
£83.05
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
15280484
|
Tenak™
TE21452B |
Boxes (10) |
£94.80
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
15210494
|
Tenak™
TE21456B |
Boxes (12) |
£103.00
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
15220514
|
Tenak™
TE21478B |
Boxes (13) |
£107.00
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
15290484
|
Tenak™
TE21454B |
Boxes (11) |
£98.70
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
Description
- Comes with a locking rod to secure the cryoboxes
- Measure the inside height of you freezer when choosing the right rack
- Constructed of stainless steel
- Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications
White cardboard boxes, 9 x 9 cell dividers | |
Stainless Steel | |
Chest Frseezers | |
50 mm (Box) |