missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Class B Graduated Polypropylene Cylinders with Raised Blue Graduations

Excellent for precise liquid measurement and fluid transfer in the laboratory.

Brand:  Hirschmann™

 View more versions of this product

Product Code. 13138029

  • £45.10 / Pack of 12

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Acc. to DIN 12681/ISO 6706
  • PP
  • Raised scale
  • Main point ring graduation
  • Hexagonal base
Specifications

Specifications

25 mL
DIN 12681, ISO 6706
B
22 mm
Round
Blue
170 mm
Polypropylene
Graduated cylinder, PP
Reusable
0.5 mL
Precise Liquid Measurement and Fluid Transfer in the Laboratory
Hexagonal
Graduated Measuring Cylinder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers