missing translation for 'onlineSavingsMsg'
Learn More

chlorodihydrido[bis[(2-diisopropylphosphino)ethyl]amine]iridium, 97%, Thermo Scientific Chemicals

Catalog Number p-4258892
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
791629-96-4
C16H37NP2·ClH2Ir
535.1
QPAONQONOKDMMJ-UHFFFAOYSA-M
iridium 3+ bis 2-diisopropylphosphanyl ethyl amine chloride dihydride, iridium 3+ chloride hydride-2-di propan-2-yl phosphanyl-n-2-di propan-2-yl phosphanyl ethyl ethan-1-amine 1/1/2/1
This item is not returnable. View return policy
791629-96-4
C16H37NP2·ClH2Ir
535.1
QPAONQONOKDMMJ-UHFFFAOYSA-M
iridium 3+ bis 2-diisopropylphosphanyl ethyl amine chloride dihydride, iridium 3+ chloride hydride-2-di propan-2-yl phosphanyl-n-2-di propan-2-yl phosphanyl ethyl ethan-1-amine 1/1/2/1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Melting Point 240.6°C to 241.9°C
Color Tan to White
Elemental Analysis 34.50 to 37.30% C, 2.50 to 2.70% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 250 mg
Formula Weight 535.1
Percent Purity 97%
Physical Form Powder
Chemical Name or Material chlorodihydrido[bis[(2-diisopropylphosphino)ethyl]amine]iridium
Show More Show Less
compliance-icons
Product Identifier
  • Chlorodihydrido(bis((2-diisopropylphosphino)ethyl)amine)iridium(III)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'