Learn More
Cole-Parmer™ Check Valves with Pipe Thread Connections
Designed for use with Cole-Parmer tubing
£144.16 - £296.00
Specifications
Product Type Check Valve
View More Specs
Products 3
Product Code Brand Maximum Pressure Price Quantity & Availability  
Product Code Brand Maximum Pressure Price Quantity & Availability  
11829462
View Documents
Cole-Parmer™
01350-24
150 psi This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11819462
View Documents
Cole-Parmer™
01350-26
125 psi This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11839462
View Documents
Cole-Parmer™
01350-30
100 psi
£296.00
Each
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
Description
Description
  • Check valve
  • With pipe thread connections
  • Available with 1/4 or 1/2 in. NPT(F) Connections
Specifications
Specifications
Check Valve